Merck
BAF148480001
Fillpour® Dialysis Tube Funnels
13.5, 19 and 28 mm Diameter Steps, 10 pak of
Größe auswählen
10 EA
97,20 €
97,20 €
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EA
97,20 €
About This Item
UNSPSC-Code:
41123100
NACRES:
NB.22
97,20 €
Check Cart for Availability
Materialien
polypropylene
Hersteller/Markenname
Bel-Art F14848-0001
Verpackung
10 pak of
Allgemeine Beschreibung
To use, cut dialysis tubing to length and knot one end. Slip the open end onto the tube funnel and secure with included rubber band or Scienceware® silicone rubber loops. A captive snap on cover has a small writing area for easy identification. Steam autoclavable at 121 deg C (250 deg F). 10 per bag.
Supports dialysis tubes on edge of vessel and allows easy filling.
Leistungsmerkmale und Vorteile
  • Durable polypropylene construction
  • Allows convenient handling of dialysis bags
  • Three tube steps fit various sizes of dialysis tubing
  • Hanger has three different notches to fit the edge of any vessel
Rechtliche Hinweise
Fillpour is a registered trademark of SP Scienceware, Division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)