Merck
Z186740
Gasbeutel
capacity, 1 gal
Synonym(e):
gas collection bag, gas collection vessel, gas sampling equipment
About This Item
UNSPSC-Code:
24111500
NACRES:
NB.51
Leistungsmerkmale
capacity 1 gal
Verpackung
pack of 1 ea
Allgemeine Beschreibung
Gas bags made of heavy natural rubber are suitable for collecting and storing large samples of gases. They are commonly used with PVC stopcocks such as Z186767.