Merck
BR759700
BRAND® stand for disposal bags
includes pack of 100 bags, PP
Synonym(e):
Accessories, Disposable Bags, Stands
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.35
Materialien
epoxy-coated (steel wire, with rubber feet)
Leistungsmerkmale
includes pack of 100 bags, PP
Verpackung
pack of 100 bags
Hersteller/Markenname
BRAND 759700
Allgemeine Beschreibung
Bags must be open during sterilization. Autoclavable (121°C)
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)