Hlavní dokumenty
SU860091
Supelco
Assembled screw cap with hole and TFE/rubber septum
black polypropylene hole cap, thread for 8-425, PTFE/rubber septum, for use with 11.6 × 32mm vial (standard opening), pkg of 100 ea
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
PTFE/rubber septum
black polypropylene hole cap
feature
closure type black polypropylene hole cap
packaging
pkg of 100 ea
septum thickness
1.3 mm
fitting
thread for 8-425
compatibility
for use with 11.6 × 32mm vial (standard opening)
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)