Hlavní dokumenty
66841-U
Supelco
Guard Cartridge Holder for BIOshell Guard Column
About This Item
UNSPSC Code:
41115700
NACRES:
SB.52
material
stainless steel holder (guard column)
Quality Level
product line
BIOshell
packaging
1 ea of
technique(s)
HPLC: suitable
General description
This Guard Cartridge Holder can be used with 4.6 mm, 3.0 mm and 2.1 mm ID BIOshell columns.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.
Navštívit knihovnu dokumentů
Sortimentní položky
BIOshell™ A400 Protein C4 columns contain 3.4 μm particles with 400 Å pores, which are derivatized with dimethylbutyl silane and exhaustively endcapped for optimum protein recovery.