Key Documents
56720-U
Supelco
Stainless Steel 1/16 in. Capillary Tubing
L × O.D. × I.D. 10 cm × 1/16 in. × 0.010 in.
About This Item
UNSPSC Code:
41115700
eCl@ss:
32119290
NACRES:
SB.52
L × O.D. × I.D.
10 cm × 1/16 in. × 0.010 in.
color
blue band
General description
316 stainless steel, 1/16 in. O.D. 1 m length is coiled, all other dimensions are straight.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)