Key Documents
29621-U
Syringes for Shimadzu AOC14, 17, 20 and 20i Autosamplers
volume 10 μL, 10R-S-GT-0.63, needle size 23 ga, needle L 42 mm
About This Item
UNSPSC Code:
41122001
NACRES:
NB.03
material
cone tip stainless steel needle
needle size
23 ga
description
color (dark orange)
feature
needle type R (removable needle)
point style 1
packaging
pkg of 1 ea
manufacturer/tradename
SGE 002902
needle L
42 mm
volume
10 μL
Legal Information
Shimadzu is a trademark of Shimadzu Corporation
related product
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)