Hlavní dokumenty
Z513547
Sigma-Aldrich
Aldrich® mini gas regulator
stainless steel body
About This Item
UNSPSC Code:
41100000
NACRES:
NB.51
material
stainless steel body
General description
These high-purity, single-stage, single-gauge regulators have a 316 SS diaphragm for accurate, stable pressure control of Aldrich liquefied gases. Delivery pressure: 1-50 psi, 1/4 in. NPTF inlet, hose barb outlet.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)