Key Documents
22085-U
Swagelok® Connector to Male NPT
stainless steel, 1/4 in. Swagelok, 1/8 in. male NPT, 400-1-2
About This Item
UNSPSC Code:
41112500
eCl@ss:
32035302
NACRES:
SB.54
material
stainless steel
packaging
pkg of 1 ea
manufacturer/tradename
400-1-2
fitting
1/4 in. Swagelok
1/8 in. male NPT
General description
Nuts and ferrules included.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)