Key Documents
Z765678
Tube rack for Benchmixer XL multi-tube vortexer
3 x 96 x 0.2 mL tube/strip carrier
Synonyma:
multi tube vortexer rack, vortexer rack
About This Item
UNSPSC Code:
41122804
NACRES:
NB.77
feature
3 x 96 x 0.2 mL tube/strip carrier
packaging
pkg of 1 ea
manufacturer/tradename
Benchmark Scientific BV1010-MP02
Legal Information
Benchmixer is a trademark of Benchmark Scientific, Inc.