Z719544
Sorenson™ low binding E tips
maximum volume 10 μL, pack of 1000 ea (Bag), non-sterile
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
volume:
10 μL
sterility:
non-sterile
feature:
barrier: no
packaging:
pack of 1000 ea (Bag)
case of 10000 (10 bags of 1000 tips)
case of 10000 (10 bags of 1000 tips)
manufacturer/tradename:
Sorenson 13980T
Doporučené produkty
material
colorless
polypropylene
sterility
non-sterile
feature
barrier: no
packaging
pack of 1000 ea (Bag)
case of 10000 (10 bags of 1000 tips)
manufacturer/tradename
Sorenson 13980T
maximum volume
10 μL
volume
10 μL
Související kategorie
General description
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Extended length. Fits Eppendorf ultra-micro pipettors.
Legal Information
Sorenson is a trademark of Sorenson BioScience, Inc.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)