Hlavní dokumenty
Z719625
Sorenson low binding standard tips
maximum volume 300 μL, case of 1000 ea (Bag), non-sterile
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
maximum volume:
300 μL
sterility:
non-sterile
feature:
barrier: no
graduations
packaging:
case of 1000 ea (Bag)
case of 10000 ea (10 bags of 1000 tips)
manufacturer/tradename:
Sorenson 31770T
material
colorless
polypropylene
sterility
non-sterile
feature
barrier: no
graduations
packaging
case of 1000 ea (Bag)
case of 10000 ea (10 bags of 1000 tips)
manufacturer/tradename
Sorenson 31770T
maximum volume
300 μL
General description
Tips feature a new surface technology designed for pipetting substances which tend to stick to the walls of pipette tips or for pipetting expensive reagents. Universal fit for research-grade pipettors.
Legal Information
Sorenson is a trademark of Sorenson BioScience, Inc.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)