Z601594
Hettich® centrifuge adapters
for ROTINA 38/38R and ROTINA 420/420R Centrifuges, Adapter for 50 mL Falcon tubes (29x115 mm)
About This Item
UNSPSC Code:
41103911
NACRES:
NB.22
Doporučené produkty
feature
Holds 50 mL tubes
packaging
pack of 4 ea
manufacturer/tradename
Hettich 1384
technique(s)
centrifugation: suitable
compatibility
for use with 1481 1490, 1495 (buckets)
General description
- compatible with bucket/carriers 1381 (Z601543) and 1430 (Z729132)
Legal Information
Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)