GE29-0220-94
ÄKTA® start protein purification system
AC/DC input 100 - 240 V AC (US and schuko plugs)
About This Item
UNSPSC Code:
41105300
NACRES:
NA.55
Doporučené produkty
description
2 buffer inlets
manufacturer/tradename
Cytiva 29-0220-94
parameter
(single wave length)
0.5 Mpa max. pressure (operating)
0.5 Mpa max. pressure (operating, sample pump)
0.5-5 mL/min flow rate
pathlength
2 mm , flow cell
viscosity
0.6-5 cP
General description
ÄKTA® start is a preparative chromatography system for laboratory scale protein purification. ÄKTA start is designed as a stand-alone system, with intuitive design, simple flow path, and user-friendly interface. With ÄKTA start, it is easy to purify a wide variety of proteins using built-in quick start methods or predefined templates, or by creating your own methods.
ÄKTA start can be combined with the Frac30 fraction collector, the user-friendly UNICORN® start control software, and application-focused prepacked columns for an automated solution.
With ÄKTA start, protein purification is quick, reliable, and easy. The system is easy to learn and use, and has an intuitive design, front-facing work area, visible flow path, and a user-friendly interface. ÄKTA start makes it easy to purify proteins, with predefined quick start and editable method templates, real-time run control and visualization, and single-click operations with a compact and portable size suitable for cold-room operations. With proven and reliable technology like LED-based UV detection, and built-in help and diagnostics, you can trust ÄKTA start to help you make an effortless switch from manual to automated protein purification.
ÄKTA start can be combined with the Frac30 fraction collector, the user-friendly UNICORN® start control software, and application-focused prepacked columns for an automated solution.
With ÄKTA start, protein purification is quick, reliable, and easy. The system is easy to learn and use, and has an intuitive design, front-facing work area, visible flow path, and a user-friendly interface. ÄKTA start makes it easy to purify proteins, with predefined quick start and editable method templates, real-time run control and visualization, and single-click operations with a compact and portable size suitable for cold-room operations. With proven and reliable technology like LED-based UV detection, and built-in help and diagnostics, you can trust ÄKTA start to help you make an effortless switch from manual to automated protein purification.
Features and Benefits
- Compact solution for quick and reliable one-step protein purification
- Sample injection and fraction collection options
- Quick start methods and templates for common purification techniques
- Intuitive touchscreen display and real-time monitoring
Other Notes
This material does not include fraction collector and software.
Legal Information
Unicorn is a registered trademark of Cytiva
ÄKTA is a registered trademark of Cytiva
recommended
Č. produktu
Popis
Stanovení ceny
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)