Key Documents
EP5810753007
Eppendorf® Adapter for 250 mL Rectangular Bucket
set of 2 pieces
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
Holds 12 x 3-15 mL tubes
packaging
set of 2 pieces
manufacturer/tradename
Eppendorf® 5810753007
technique(s)
centrifugation: suitable
compatibility
for use with 250 mL rectangular buckets
Legal Information
Eppendorf is a registered trademark of Eppendorf AG