Key Documents
EP5810752000
Eppendorf® Adapter for 250 mL Rectangular Bucket
Holds 15 x 2,6-7 mL tubes, set of 2 pieces
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
Holds 15 x 2,6-7 mL tubes
packaging
set of 2 pieces
manufacturer/tradename
Eppendorf® 5810752000
technique(s)
centrifugation: suitable
compatibility
for use with 250 mL rectangular buckets
Legal Information
Eppendorf is a registered trademark of Eppendorf AG