EP5804757009
Eppendorf® Adapter For 100 mL Rectangular Bucket
holds 18-30 mL tubes, pack of 2 ea
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
Doporučené produkty
feature
holds 18-30 mL tubes
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5804757009
technique(s)
centrifugation: suitable
compatibility
for use with 100 mL rectangular bucket
General description
Tube adapters for use with Eppendorf 100 mL rectangular bucket.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)