Hlavní dokumenty
EP5804715004
Eppendorf® Fixed-angle rotor
F45-30-11 30 x 1.5/2 ml, incl. aluminium lid
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 30 x 1.5/2.0 mL tubes
F45-30-11 30 x 1.5/2 ml, incl. aluminium lid
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5804715004
parameter
14,000 max. RCF (20,817 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
General description
Fixed angle rotors for use with Eppendorf centrifuge models.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)