Key Documents
EP5804726006
Eppendorf® Fixed-angle rotor
FA-45-30-11, 30x 1,5/2 ml, incl. aluminium lid, aerosol-tight
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 30 x 1.5/2.0 mL tubes
FA-45-30-11, 30x 1,5/2 ml, incl. aluminium lid, aerosol-tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5804726006
parameter
14,000 max. RCF (20,817 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5804/5804R/5810/5810R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG