EP5702737003
Eppendorf® Adapter for 85/100 mL Round Bucket
holds Vacutainer and Falcon tubes 2-7 ml, pack of 2 ea
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
Doporučené produkty
feature
holds Vacutainer and Falcon tubes 2-7 ml
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5702737003
technique(s)
centrifugation: suitable
compatibility
for use with 100 mL round buckets
General description
adapter for use with Eppendorf 85/100 mL round bucket
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)