Z258431
Head set for Vortex-Genie® 2
Multiple head
About This Item
UNSPSC Code:
41103814
NACRES:
NB.73
Doporučené produkty
manufacturer/tradename
Scientific Industries
General description
Complete with 6in. platform head, one microtube insert, and one 96 well plate insert.
Legal Information
Vortex-Genie is a registered trademark of Scientific Industries
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)