Key Documents
EP5702706000
Eppendorf® 10 adapters 15 mL Falcon tubes
Holds 15 mL tubes
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
Holds 15 mL tubes
packaging
pack of 10 ea
manufacturer/tradename
Eppendorf® 5702706000
technique(s)
centrifugation: suitable
compatibility
for use with Rotor F-35-30-17
Legal Information
Eppendorf is a registered trademark of Eppendorf AG