Hlavní dokumenty
EP5495503005
Rotor for Eppendorf® 5425 Centrifuge
FA-24x2-PTFE, with PTFE coating, to hold, 24 x 1.5/2.0 mL tubes, lid
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
lid
to hold 24 x 1.5/2.0 mL tubes
FA-24x2-PTFE
with PTFE coating
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5495503005
parameter
15,060 max. RCF (21,300 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5425
General description
Rotor for Eppendorf(R) 5425 Centrifuge
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)