Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 14 900,00 Kč |
O této položce
14 900,00 Kč
feature
lid, to hold 18 x 1.5/2.0 mL tubes or spin columns, FA-18x2-Kit
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5495508007
parameter
15,060 max. RCF (18,565 xg)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5425
General description
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description FA-18x2-Kit, to hold, 18 x 1.5/2.0 mL tubes or spin columns, lid | description FA-24x2, lid | description FA-24x2-PTFE, with PTFE coating, to hold, 24 x 1.5/2.0 mL tubes, lid | description FA-10x5, to hold, 10 x 5.0 mL Eppendorf tubes, lid |
| compatibility for use with centrifuge models 5425 | compatibility for use with centrifuge models 5425 | compatibility for use with centrifuge models 5425 | compatibility for use with centrifuge models 5425 |
| manufacturer/tradename Eppendorf® 5495508007 | manufacturer/tradename Eppendorf® 5495500006 | manufacturer/tradename Eppendorf® 5495503005 | manufacturer/tradename Eppendorf® 5495505008 |
| feature FA-18x2-Kit, to hold 18 x 1.5/2.0 mL tubes or spin columns, lid | feature lid, FA-24x2 | feature lid, to hold 24 x 1.5/2.0 mL tubes, with PTFE coating, FA-24x2-PTFE | feature FA-10x5, lid, to hold 10 x 5.0 mL Eppendorf tubes |
| parameter 15,060 max. RCF (18,565 xg) | parameter 15,060 max. RCF (21,300 xg) | parameter 15,060 max. RCF (21,300 xg) | parameter 15,060 max. RCF (21,300 xg) |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| technique(s) centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) centrifugation: suitable |
Still not finding the right product?
Explore all of our products under Rotor for Eppendorf® 5425 Centrifuge
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?