Key Documents
EP5490038002
Rotor for Eppendorf® Concentrator Plus
F-45-8-17, non aerosol tight, to hold, 8 x 15 mL conical tubes
About This Item
UNSPSC Code:
41103909
NACRES:
NB.82
feature
to hold 8 x 15 mL conical tubes
to hold 8 x 17 x 118 - 123 mm
fixed angle rotor
non aerosol tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5490038002 (EU)
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models Concentrator Plus
Legal Information
Eppendorf is a registered trademark of Eppendorf AG