Vybrat velikost
| Vám/SKU | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 17 800,00 Kč |
O této položce
17 800,00 Kč
feature
lid, to hold 30 x 1.5/2.0 mL tubes, aerosol tight, fixed angle rotor
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 5409706004 (Global)
parameter
14,000 rpm, 20,800 × g max. RCF
technique(s)
centrifugation: suitable
compatibility
for use with centrifuge models 5427R
General description
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| description FA-45-30-11, aerosol tight, to hold, 30 x 1.5/2.0 mL tubes, lid | description FA-45-30-11, non aerosol-tight, to hold, 30 x 1.5/2.0 mL tubes, with rotor lid | description FA-45-24-11, aerosol tight, to hold, 24 x 1.5/2.0 mL tubes, lid | description FA-45-48-11, aerosol-tight, to hold, 48 x 1.5/2.0 mL tubes, lid |
| compatibility for use with centrifuge models 5427R | compatibility for use with centrifuge models 5427R | compatibility for use with centrifuge models 5427R | compatibility for use with centrifuge models 5427R |
| manufacturer/tradename Eppendorf® 5409706004 (Global) | manufacturer/tradename Eppendorf® 5409708007 (Global) | manufacturer/tradename Eppendorf® 5409702009 (Global) | manufacturer/tradename Eppendorf® 5409710001 (Global) |
| feature lid, aerosol tight, to hold 30 x 1.5/2.0 mL tubes, fixed angle rotor | feature fixed angle rotor, to hold 30 x 1.5/2.0 mL tubes, non aerosol-tight, with rotor lid | feature aerosol tight, fixed angle rotor, to hold 24 x 1.5/2.0 mL tubes, lid | feature lid, to hold 48 x 1.5/2.0 mL tubes, fixed angle rotor, aerosol-tight |
| parameter 14,000 rpm, 20,800 × g max. RCF | parameter 14,000 rpm, 20,800 × g max. RCF | parameter 25,000 × g max. RCF, 16,220 rpm | parameter 18,210 × g max. RCF, 12,700 rpm |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| technique(s) centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) centrifugation: suitable | technique(s) centrifugation: suitable |
Still not finding the right product?
Explore all of our products under Rotor for Eppendorf® 5427R Centrifuge
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?