Key Documents
EP5427743006
Adapter for Eppendorf® F-35-6-30 Rotor
holds 1 x 90 - 110 mm tube, large rotor bore, large rotor bore, pack of 2 ea
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
holds 1 x 90 - 110 mm tube, large rotor bore
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5427743006 (Global)
parameter
7,129 × g max. RCF
technique(s)
centrifugation: suitable
diam.
13 mm
compatibility
for use with Rotor F-35-6-30 (large rotor bore)
General description
Adapters to fit various size tubes and bottles for use with Eppendorf rotor F-35-6-30.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG