Key Documents
EP5427742000
Adapter for Eppendorf® F-35-6-30 Rotor
holds 1 x 65 - 89 mm tube, large rotor bore, large rotor bore, pack of 2 ea
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
holds 1 x 65 - 89 mm tube, large rotor bore
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5427742000 (Global)
parameter
6,100 × g max. RCF
technique(s)
centrifugation: suitable
diam.
13 mm
compatibility
for use with Rotor F-35-6-30 (large rotor bore)
General description
Adapters to fit various size tubes and bottles for use with Eppendorf rotor F-35-6-30.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG