Key Documents
EP5427704000
Eppendorf® Rotor Lid
for FA-45-24-11-Kit, aerosol-tight
About This Item
UNSPSC Code:
41103913
NACRES:
NB.82
feature
for FA-45-24-11-Kit, aerosol-tight
manufacturer/tradename
5427704000
Eppendorf® 5427704000
Eppendorf®
technique(s)
centrifugation: suitable
compatibility
for use with 5430 R
for use with 5430
Legal Information
Eppendorf is a registered trademark of Eppendorf AG