Key Documents
EP5409703005
Lid for Eppendorf(R) FA-45-24-11 Rotor
aerosol-tight, for 5427R, aluminum
About This Item
UNSPSC Code:
41103913
NACRES:
NB.82
material
aluminum
feature
aerosol-tight
manufacturer/tradename
Eppendorf® 5409703005 (Global)
technique(s)
centrifugation: suitable
compatibility
for use with 5427 R
Legal Information
Eppendorf is a registered trademark of Eppendorf AG