Key Documents
26159
Hamilton® plunger assemblies with tips for series 1000, 1700, and 1800
for use with Hamilton 1001 LT-N-LTN-RN, pkg of 1 ea
About This Item
UNSPSC Code:
41122001
material
PTFE plunger tip
size
1 mL
feature
1001 LT-N-LTN-RN
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 1359-01
compatibility
for use with Hamilton 1001 LT-N-LTN-RN
General description
For field repair of Hamilton Series 1000, 1700 and 1800 syringes.
Application
Plunger assemblies has been used during constructing a mixer which was for rapid and convenient hydration in the determination of small viscous lipid-containing samples.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)
Lot/Batch Number
Je nám líto, ale pro tento produkt momentálně nemáme COA k dispozici online.
Navštívit knihovnu dokumentů
A simple mechanical mixer for small viscous lipid-containing samples.
Cheng, Anchi, et al.
Chemistry and Physics of Lipids, 95.1, 11-21 (1998)