Vybrat velikost
| Vám/Skladová položka | Dostupnost | Cena |
|---|---|---|
1 ea | Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti. | 1 620,00 Kč |
O této položce
1 620,00 Kč
material
PTFE Luer lock
needle size
(not included)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 81320
needle L
, n/a
volume
1 mL
fitting
luer lock (without slots)
compatibility
for use with Hamilton 1001TLL (PTFE Luer lock)
General description
Application
Legal Information
1 of 1
Tato položka | |||
|---|---|---|---|
| material PTFE Luer lock | material PTFE Luer lock | material PTFE Luer lock (with slots) | material Kel-F CTFE Luer lock |
| fitting luer lock (without slots) | fitting luer lock (without slots) | fitting luer lock (without slots) | fitting luer lock (without slots) |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| manufacturer/tradename Hamilton 81320 | manufacturer/tradename Hamilton 81520 | manufacturer/tradename Hamilton 81327 | manufacturer/tradename Hamilton 86020 |
| needle L , n/a | needle L , n/a | needle L , n/a | needle L , n/a |
| volume 1 mL | volume 5 mL | volume 1 mL | volume 100 mL |
Still not finding the right product?
Explore all of our products under Hamilton® syringe, 1000 series GASTIGHT®, PTFE luer lock
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?