Key Documents
1.03831
Supelco
HPLC bottle adapter S40 with 3 tube connections and 1 connection for exhaust air filter, solvents disposal
About This Item
UNSPSC Code:
41121516
NACRES:
NA.03
storage temp.
2-30°C
Související kategorie
General description
Safe, easy and contamination free solvent handling is one of the main important topics in daily lab work. Our bottle adapter with included filter-valve connects your S40-threaded glass bottles for solvents disposal directly to instruments with 3mm tubes. This avoids escaping solvent vapours and other harmful gases to protect your health and the environment. The specially selected materials provide highest safety and maximum protection against solvent contact The adapter for disposal bottles has to be completed by the appropriate exhaust air filter 1.03833.0001.