Key Documents
Z553875
Sigma-Aldrich
Aldrich® safety bubbler with SafetyBarb® hose connectors
capacity 15 mL , fill mark
Synonyma:
bubblers, safety bubblers, scientific bubblers
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
capacity
15 mL , fill mark
Související kategorie
General description
Built-in flash arrester bulb prevents backflow. Order replacement SafetyBarb hose connectors Z547786 (straight) or Z547883 (angled).
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany