Merck
Z741751
3-cylinder bubble trap
Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
O této položce
NACRES:
NB.22
UNSPSC Code:
41121800
eCl@ss:
32150313
Material:
polycarbonate
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
material
polycarbonate
sterility
sterile; irradiated
feature
autoclavable: no
manufacturer/tradename
IBI ACCFL0002
General description
The IBI 3-cylinder bubble trap is designed to be used in conjunction with the IBI flow cell systems. It comes complete with the FLCAS0001 3-channel flow cell assembly and can be purchased separately and used with other IBI flow cell components.
The 3-cylinder bubble trap is made of polycarbonate and is a single use device that comes irradiated (sterilized) and ready for use. Its primary function is to remove air bubbles from the tubing of a flow cell system before they reach the growth cell. The bubble trap also helps eliminate pulsations that can occur in the flow due to peristaltic pump action.
The 3-cylinder bubble trap will accommodate one, 3-channel flow cell or three, convertible flow cells. Since each cylinder is separate, not all cylinders need to be connected to the flow cell system.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)