Z530352
Aldrich® reaction vessel bubbler
joint: ST/NS 29/32
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Doporučené produkty
joint
joint: ST/NS29/32
capacity
5-7 mL
General description
Excellent for closing off a reaction vessel from the atmosphere.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)