Key Documents
Z515965
Sigma-Aldrich
Aldrich® solvent storage flask
capacity 1,000 mL, joint: ST/NS 24/40
Synonyma:
bottom flask, flask round, round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
round bottom
single-neck
joint
joint: ST/NS24/40
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
1,000 mL
General description
With ground-glass joint and glass stopcock. Use septum Z565695 or Z565679 (see Septa Section).