Hlavní dokumenty
Z513334
Micro spatulas
flat rounded/tapered PTFE coated ends, L 7 5/8 in. (190 mm)
About This Item
UNSPSC Code:
41122403
NACRES:
NB.35
material
nickel-stainless steel
packaging
pkg of 1 ea
L
7 5/8 in. (190 mm)
General description
Nickel stainless, PTFE coated spatula with one rounded end and one tapered end. Rounded end approximately 5/16 in. (8 mm) wide x 2 in. (51 mm) long. Tapered end approximately 1/4 in. (6 mm) wide x 1 7/8 in. (48 mm) long.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)