Key Documents
Z310743
Volac® Pasteur pipette
Cotton plugged, L 150 mm
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
soda-lime glass
sterility
non-sterile
feature
graduations: no
Cotton plugged
packaging
pack of 1000 ea
pack of 1000
manufacturer/tradename
Volac D810/PP
availability
not available in USA and Canada
L
150 mm
Legal Information
Volac is a registered trademark of Poulten & Graf Ltd.
Zákazníci si také prohlíželi
Slide 1 of 7
1 of 7
BRAND® pipette withdraw volume 2 mL (including bulb), graduations, 0.5 mL, drop quantity 22 - 26 per mL
BR747765
BRAND® pipette
Pasteur pipettes long capillary tip, approx 2 mL withdraw volume, soda-lime glass
Z628018
Pasteur pipettes
BRAND® pipette withdraw volume 5.5 mL (including bulb), graduations, 0.25 mL
BR747775
BRAND® pipette
BRAND® pipette withdraw volume 3 mL (including bulb), graduations: no, drop quantity 25 per mL
Z331740
BRAND® pipette