Key Documents
Z286443
Nalgene® stopcock
2-way, bore size 4 mm
Synonyma:
2-way connector, hose connector
About This Item
UNSPSC Code:
41100000
NACRES:
NB.79
material
polypropylene (with TFE plug)
description
for tubing I.D. 1/4 -5/16 in.
feature
autoclavable
packaging
pack of 1 ea
case of 6 ea
manufacturer/tradename
Nalgene® 6460, 0004
bore size
4 mm
General description
Serrated tubulation on each end.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Zákazníci si také prohlíželi
Slide 1 of 5
1 of 5