Hlavní dokumenty
Z223670
Sigma-Aldrich
Aldrich® septum-inlet adapter, in-line
Plain, Joints: ST/NS 29/32 (2)
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
joint
Joints: ST/NS29/32 (2)
General description
Septum-inlets require septum Z565695 or Z565679 except where noted.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)