Key Documents
Z223220
Sigma-Aldrich
Aldrich® in-line oil bubbler
joint: ST/NS 14/20
Synonyma:
bubblers, in-line bubblers, scientific bubblers
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
joint
joint: ST/NS14/20
capacity
10 mL
Související kategorie
General description
For precise N2 pressure control during inert atmosphere reactions. Connect reaction vessel to in-line joint or use with a ballast bulb to keep pressure constant.