Key Documents
Z204161
Supelco
Rectangular TLC developing tanks, complete
L × H × W 17.5 cm × 16.0 cm × 6.2 cm
About This Item
UNSPSC Code:
41115711
NACRES:
NC.07
technique(s)
thin layer chromatography (TLC): suitable
L × H × W
17.5 cm × 16.0 cm × 6.2 cm
Footnote
TLC Developing Tanks come complete with the developing tank and lid