Hlavní dokumenty
Z156477
Sigma-Aldrich
O-ring set
nitrile
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
material
nitrile
manufacturer/tradename
Ace Glass
General description
500 O-rings in a storage box. Thirty sizes ranging from: 1/8 I.D. × 1/4 in. O.D. up to 1 3/4 I.D. × 2 1/8 in. O.D. Use nitrile for economical sealing, Viton® for thermal or chemical resistance.
Legal Information
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)