Z101915
Single-bulb ball-tube
capacity 250 mL, joint: ST/NS 24/40, plain
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
Doporučené produkty
joint
joint: ST/NS24/40
capacity
250 mL
General description
Arranged horizontally outside of Kugelrohr oven, the ball-tubes cool and collect distillate with minimal loss of material.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)