Key Documents
HAM59000-05
Hamilton® Microlab® 600 instrument syringe
model, 1701 BFP, volume 10 μL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
Gastight Borosilicate glass/PTFE syringe
feature
model 1701 BFP
manufacturer/tradename
Hamilton 59000-05
parameter
10-115 °C temp. range (50-239 °F)
1000 psig pressure
I.D. × O.D.
0.460 mm × 6.60 mm
volume
10 μL
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Microlab is a registered trademark of Hamilton Co.