DWK73802-22400
KIMBLE® Solid Cap
black phenolic cap (with rubber liner), 22-400 thread
About This Item
UNSPSC Code:
41121800
NACRES:
NB.11
Doporučené produkty
material
PTFE/rubber liner
black phenolic cap (with rubber liner)
description
with rubber liner, phenolic
sterility
non-sterile
feature
class A
packaging
case of 144 ea
manufacturer/tradename
Kimble® 73802-22400
fitting
22-400 thread
Související kategorie
General description
Class A Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
- Specially formulated phenolic cap material
- Autoclavable
- White rubber liners
- Excellent for general laboratory use
Features and Benefits
- Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving
- PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
- Excellent for general laboratory use
- Identified in ASTM Specification E982, Class A requirements
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)