Key Documents
BR6735
Filling valve with for Dispensette® S
25, 50 and 100 mL, PFA/glass/ceramic
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
manufacturer/tradename
BRAND 6735
packaging
1 ea of
capacity
25, 50 and 100 mL
General description
Valve: PFA/ETFE/Boro 3.3/ceramic.
Legal Information
Dispensette is a registered trademark of BRAND GMBH + CO KG