Z653020
Julabo® bath test tube racks
to fit 90 test tubes, for open bath circulators, diam. 12-13 mm, polypropylene
About This Item
UNSPSC Code:
41122804
NACRES:
NB.77
Doporučené produkty
material
polypropylene
feature
Holds 90 x 12-13 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Julabo 8970306
parameter
+80 °C temp. limit
diam.
12-13 mm
Legal Information
Julabo is a registered trademark of Julabo USA, Inc.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)