Hlavní dokumenty
924598
Sigma-Aldrich
Cross-shaped channel chip (slide format)
Fluidic 160, Cyclic Olefin Copolymer (COC) with Mini-Luer Interface
About This Item
UNSPSC Code:
42142600
NACRES:
NA.23
Application
This is a Cross-shaped channel chip in a microscopy slide format. These chips offer two separate channel structures with crossing channels on each device. One of those with, one without a channel offset. The microfluidic chips of this family feature Mini Luer interfaces.
Cross-shaped channel chip (slide format), Fluidic 160, is made of COC (Cyclic Olefin Copolymer) with Mini-Luer Interface.
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.
Navštívit knihovnu dokumentů
Recent advances of controlled drug delivery using microfluidic platforms.
Sanjay, et al.
Advanced Drug Delivery Reviews, 128, 3-28 (2020)
Dongfei Liu et al.
Lab on a chip, 17(11), 1856-1883 (2017-05-10)
The microfluidic technique has brought unique opportunities toward the full control over the production processes for drug delivery carriers, owing to the miniaturisation of the fluidic environment. In comparison to the conventional batch methods, the microfluidic setup provides a range
Sharma T Sanjay et al.
Advanced drug delivery reviews, 128, 3-28 (2017-09-19)
Conventional systematically-administered drugs distribute evenly throughout the body, get degraded and excreted rapidly while crossing many biological barriers, leaving minimum amounts of the drugs at pathological sites. Controlled drug delivery aims to deliver drugs to the target sites at desired